Boric acid, ACS, 99.5% min, Thermo Scientific Chemicals
Boric acid, ACS, 99.5% min, Thermo Scientific Chemicals
Boric acid, ACS, 99.5% min, Thermo Scientific Chemicals
Boric acid, ACS, 99.5% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Boric acid, ACS, 99.5% min, Thermo Scientific Chemicals
In hardening steel, preservatives, manufacturing of cements, porcelain, enamels, glass, borates
Have Questions?
Catalog NumberQuantity
ALF033253.36500 g
Catalog number ALF033253.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS100-14-1
IUPAC Name1-(chloromethyl)-4-nitrobenzene
Molecular FormulaC7H6ClNO2
InChI KeyKGCNHWXDPDPSBV-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(CCl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Crystalline powder or needles
Melting point71°C to 74°C
In hardening steel, preservatives, manufacturing of cements, porcelain, enamels, glass, borates, artificial gems, in nickeling baths, painting, photographyIt is a precursor material for other boron compounds. A dilute water solution of boric acid is usually employed as a mild antiseptic and eyewash. Boric acid is too employed in leather manufacture, electroplating, and cosmetics.