Tripotassium citrate monohydrate, 99+%, Thermo Scientific Chemicals
Tripotassium citrate monohydrate, 99+%, Thermo Scientific Chemicals
Tripotassium citrate monohydrate, 99+%, Thermo Scientific Chemicals
Tripotassium citrate monohydrate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tripotassium citrate monohydrate, 99+%, Thermo Scientific Chemicals
CAS: 6100-05-6 | C6H7K3O8 | 324.41 g/mol
Have Questions?
Catalog NumberQuantity
ALF033222.36500 g
Catalog number ALF033222.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTripotassium citrate monohydrate
CAS6100-05-6
Melting Point230°C (decomposition)
Recommended StorageAmbient temperatures
Density1.98
View more
Tripotassium citrate hydrate is used in food and beverage industries. It finds application as buffering, sequestering or emulsifying agent. It is also used as a potassium source in pharmaceuticals and an active ingredient to treat urinary duct stones. It is added to oral care products to reduce the teeth sensitivity.