Citric acid diammonium salt, 99+%, Thermo Scientific Chemicals
Citric acid diammonium salt, 99+%, Thermo Scientific Chemicals
Citric acid diammonium salt, 99+%, Thermo Scientific Chemicals
Citric acid diammonium salt, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Citric acid diammonium salt, 99+%, Thermo Scientific Chemicals
CAS: 3012-65-5 | C6H14N2O7 | 226.19 g/mol
Have Questions?
Catalog NumberQuantity
ALF033221.36500 g
Catalog number ALF033221.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS629-09-4
IUPAC Name1,6-diiodohexane
Molecular FormulaC6H12I2
InChI KeyQLIMAARBRDAYGQ-UHFFFAOYSA-N
SMILESICCCCCCI
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5840-1.5890 @ 20?C
Appearance (Color)Clear colorless to yellow to brown
Assay (GC)≥97.0%
Identification (FTIR)Conforms
Citric acid diammonium salt is used in pharmaceuticals, rust proofing, cotton printing and plasticizers. It is used as a reagent for the determination of phosphate in fertilizers, and as a buffering agent, chelating agent and source of nitrogen in chicken food supplements.