Phenol, ACS, 99+%, stab., Thermo Scientific Chemicals
Phenol, ACS, 99+%, stab., Thermo Scientific Chemicals
Phenol, ACS, 99+%, stab., Thermo Scientific Chemicals
Phenol, ACS, 99+%, stab., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phenol, ACS, 99+%, stab., Thermo Scientific Chemicals
CAS: 108-95-2 | C6H6O | 94.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number ALF033213.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
GradeACS
Unit Size
Phenol is used as a precursor for cyclohexanone, plastics, nonionic detergents and pharmaceutical drugs like aspirin. It acts as an anesthetic in chloraseptic. It reacts with acetone to get bisphenol-A which is used in the synthesis of poly carbonates and epoxide resins. It is also used in the manufacture of synthetic resins, dyes, pharmaceuticals, synthetic tanning agents, perfumes, lubricating oils and solvents. In molecular biology, it is used in the extraction of nucleic acid from tissues by using the liquid/liquid phenol-chloroform extraction technique. It is an active component of paint strippers which are used for the removal of epoxy and polyurethane. It is also used in the preparation of cosmetics, hair colors and skin lightening preparations. In the field of medicine, its spray is useful in helping sore throat.