Chloroform, Spectrophotometric Grade, 99.5+%, Thermo Scientific Chemicals
Chloroform, Spectrophotometric Grade, 99.5+%, Thermo Scientific Chemicals
Chloroform, Spectrophotometric Grade, 99.5+%, Thermo Scientific Chemicals
Chloroform, Spectrophotometric Grade, 99.5+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chloroform, Spectrophotometric Grade, 99.5+%, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF032442.K21 L
Catalog number ALF032442.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS13939-06-5
IUPAC Namehexakis(methanidylidyneoxidanium) molybdenum
Molecular FormulaC6MoO6
InChI KeyKMKBZNSIJQWHJA-UHFFFAOYSA-N
SMILES[Mo].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Elemental Analysis>=97.5 %
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
Chloroform is used in spectrophotometry and environmental testing.