Methanol, ultrapure, Spectrophotometric Grade, 99.8+%, Thermo Scientific Chemicals
Methanol, ultrapure, Spectrophotometric Grade, 99.8+%, Thermo Scientific Chemicals
Methanol, ultrapure, Spectrophotometric Grade, 99.8+%, Thermo Scientific Chemicals
Methanol, ultrapure, Spectrophotometric Grade, 99.8+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methanol, ultrapure, Spectrophotometric Grade, 99.8+%, Thermo Scientific Chemicals
CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
Catalog number ALF032435.K7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 L
Request bulk or custom format
Chemical Identifiers
CAS1266615-56-8
IUPAC Namesodium 4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]benzen-1-olate
Molecular FormulaC12H6Cl2NNaO2
InChI KeyCVSUAFOWIXUYQA-UHFFFAOYSA-M
SMILES[Na+].[O-]C1=CC=C(C=C1)N=C1C=C(Cl)C(=O)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark green to dark blue-purple
Appearance (Form)Powder
Infrared spectrumConforms
Titration with HClO494 to 106 % (On dry substance)
Loss on drying=<14 % (1 g, 120°C)
View more
Methanol is widely used as a solvent in synthetic organic chemistry. It is also used as a solvent in analytical chemistry, especially for HPLC, LC-MS, GC-MS, ICP-MS and spectrophotometry. It is used in the production of biodiesel, formaldehyde and acetic acid. It is also used as fuel, antifreeze, denaturant for ethanol and an energy carrier for fuel cell application.