Phthalocyanine, Thermo Scientific Chemicals
Phthalocyanine, Thermo Scientific Chemicals
Phthalocyanine, Thermo Scientific Chemicals
Phthalocyanine, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phthalocyanine, Thermo Scientific Chemicals
CAS: 574-93-6 | C32H18N8 | 514.552 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number ALF032073.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS108-78-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
HPLC>=98.5 %
Infrared spectrumConforms
Pthalocyanine is a common macrocylic blue-green dye. It is also used as a suitable laser dye metal. Phthalocyanines have long been examined as catalysts for redox reactions. Areas of interest are the oxygen reduction reaction and the sweetening of gas streams by removal of hydrogen sulfide. Phthalocyanine compounds have been investigated as donor materials in molecular electronics.