Iodomethane, 99.5%, stab. with copper, Thermo Scientific Chemicals
Iodomethane, 99.5%, stab. with copper, Thermo Scientific Chemicals
Iodomethane, 99.5%, stab. with copper, Thermo Scientific Chemicals
Iodomethane, 99.5%, stab. with copper, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iodomethane, 99.5%, stab. with copper, Thermo Scientific Chemicals
CAS: 74-88-4 | CH3I | 141.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF031876.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS67-48-1
IUPAC Name(2-hydroxyethyl)trimethylazanium chloride
Molecular FormulaC5H14ClNO
InChI KeySGMZJAMFUVOLNK-UHFFFAOYSA-M
SMILES[Cl-].C[N+](C)(C)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Adhering crystals
Water=<2 %
Appearance (Color)White
Infrared spectrumConforms
Titration Argentometric98.5 to 101.5 % (on dry substance)
Iodomethane is a methylating agent used for alkylating carbon, oxygen, sulfur, nitrogen and phosphorus nucleophiles. For example, phenol reacts with methyl iodide to give anisole. In the Monsanto process, the insitu formed iodomethane reacts with carbon monoxide in the presence of a rhodium complex to give acetyl iodide, which on hydrolysis yields acetic acid. It reacts with magnesium to form the Grignard reagent, methylmagnesium iodide used in organic synthesis. It finds applications as an intermediate in the manufacture of pharmaceuticals and in phase transfer catalysts.