Iodomethane, 99.5%, stab. with copper, Thermo Scientific Chemicals
Iodomethane, 99.5%, stab. with copper, Thermo Scientific Chemicals
Iodomethane, 99.5%, stab. with copper, Thermo Scientific Chemicals
Iodomethane, 99.5%, stab. with copper, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Iodomethane, 99.5%, stab. with copper, Thermo Scientific Chemicals
CAS: 74-88-4 | CH3I | 141.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALF031876.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialIodomethane
CAS74-88-4
Health Hazard 1H301+H331-H312-H315-H319-H335-H351
Health Hazard 3P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P312-P330-P332+P313-P362-P501c
Melting Point-67°C
View more
Iodomethane is a methylating agent used for alkylating carbon, oxygen, sulfur, nitrogen and phosphorus nucleophiles. For example, phenol reacts with methyl iodide to give anisole. In the Monsanto process, the insitu formed iodomethane reacts with carbon monoxide in the presence of a rhodium complex to give acetyl iodide, which on hydrolysis yields acetic acid. It reacts with magnesium to form the Grignard reagent, methylmagnesium iodide used in organic synthesis. It finds applications as an intermediate in the manufacture of pharmaceuticals and in phase transfer catalysts.