Tris(hydroxymethyl)aminomethane, ACS, 99.8-100.1% (Assay, dried basis), Thermo Scientific Chemicals
Tris(hydroxymethyl)aminomethane, ACS, 99.8-100.1% (Assay, dried basis), Thermo Scientific Chemicals
Tris(hydroxymethyl)aminomethane, ACS, 99.8-100.1% (Assay, dried basis), Thermo Scientific Chemicals
Tris(hydroxymethyl)aminomethane, ACS, 99.8-100.1% (Assay, dried basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tris(hydroxymethyl)aminomethane, ACS, 99.8-100.1% (Assay, dried basis), Thermo Scientific Chemicals
CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol
Have Questions?
Catalog NumberQuantity
ALF031801.30250 g
Catalog number ALF031801.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS7778-18-9
IUPAC Namecalcium sulfate
Molecular FormulaCaO4S
InChI KeyOSGAYBCDTDRGGQ-UHFFFAOYSA-L
SMILES[Ca++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Water=<0.3 %
Particle size10 to 20 mesh
Appearance (Form)Irregular granules
Composition>=99.5 % (CaSO4) Typical
Tris(hydroxymethyl)aminomethane is used as a buffer in biochemistry and molecular biology laboratories. It is used as a primary standard to standardize acid solutions for chemical analysis. It finds application in cell membranes to increase permeability. As an alternative to sodium bicarbonate, it used in the treatment of metabolic acidosis. It acts as a precursor for the preparation of polymers, oxazolones and oxazolidines.