Methanol, ACS, absolute, low acetone, 99.8+%, Thermo Scientific Chemicals
Methanol, ACS, absolute, low acetone, 99.8+%, Thermo Scientific Chemicals
Methanol, ACS, absolute, low acetone, 99.8+%, Thermo Scientific Chemicals
Methanol, ACS, absolute, low acetone, 99.8+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methanol, ACS, absolute, low acetone, 99.8+%, Thermo Scientific Chemicals
CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
Catalog number ALF031721.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS7439-95-4
IUPAC Namemagnesium
Molecular FormulaMg
InChI KeyJLVVSXFLKOJNIY-UHFFFAOYSA-N
SMILES[Mg]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥99.8%
Appearance (Color)Metallic white to grey
FormPowder
Particle Size>45 µm : ≤ 8.0%
Particle Size<45 µm : ≥ 92.0%
Methanol is useful in the synthesis of nucleic acids and peptides. It is used as an antifreeze, solvent, fuel, and as a denaturant for ethanol. It is also used for producing biodiesel. It is used for the synthesis of formaldehyde and acetic acid. It is also useful to make chemicals, to remove water from automotive and aviation fuels, as a solvent for paints and plastics, and as an ingredient in a wide variety of products. It plays an important role for the measurement of methyl ester content of pectin.