Methanol, ACS, absolute, low acetone, 99.8+%, Thermo Scientific Chemicals
Methanol, ACS, absolute, low acetone, 99.8+%, Thermo Scientific Chemicals
Methanol, ACS, absolute, low acetone, 99.8+%, Thermo Scientific Chemicals
Methanol, ACS, absolute, low acetone, 99.8+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methanol, ACS, absolute, low acetone, 99.8+%, Thermo Scientific Chemicals
CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number ALF031721.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS62-56-6
IUPAC Namethiourea
Molecular FormulaCH4N2S
InChI KeyUMGDCJDMYOKAJW-UHFFFAOYSA-N
SMILESNC(N)=S
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point171°C to 177°C
Titration Iodimetric>=98.5 %
Loss on drying=<0.5 %
Appearance (Form)Crystalline powder or crystals
Sulfated ash=<0.1 %
View more
Methanol is useful in the synthesis of nucleic acids and peptides. It is used as an antifreeze, solvent, fuel, and as a denaturant for ethanol. It is also used for producing biodiesel. It is used for the synthesis of formaldehyde and acetic acid. It is also useful to make chemicals, to remove water from automotive and aviation fuels, as a solvent for paints and plastics, and as an ingredient in a wide variety of products. It plays an important role for the measurement of methyl ester content of pectin.