1,3-Diphenylguanidine, primary standard, 99+%, Thermo Scientific Chemicals
1,3-Diphenylguanidine, primary standard, 99+%, Thermo Scientific Chemicals
1,3-Diphenylguanidine, primary standard, 99+%, Thermo Scientific Chemicals
1,3-Diphenylguanidine, primary standard, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,3-Diphenylguanidine, primary standard, 99+%, Thermo Scientific Chemicals
Primary material for standardizing acids
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number ALF031395.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS151982-51-3
IUPAC Name4-bromo-2-fluorobenzoyl chloride
Molecular FormulaC7H3BrClFO
InChI KeyPCFIABOQFAFDAU-UHFFFAOYSA-N
SMILESFC1=CC(Br)=CC=C1C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline mass or liquid
GC>=98.5 %
Appearance (Color)White to light yellow
Infrared spectrumConforms
Primary material for standardizing acids. Diphenylguanidine is used as a medium of natural and synthetic rubber accelerator. It acts as a complexing agent for the detection of metals and organic bases. Further, it is used as a secondary foam stabilizer in the silicoflouride foam process. It is actively involved in the manufacture of rubber sheet, rubber shoes and other rubber industrial products.