1,5-Diphenylcarbazide, ACS, Thermo Scientific Chemicals
1,5-Diphenylcarbazide, ACS, Thermo Scientific Chemicals
1,5-Diphenylcarbazide, ACS, Thermo Scientific Chemicals
1,5-Diphenylcarbazide, ACS, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,5-Diphenylcarbazide, ACS, Thermo Scientific Chemicals
CAS: 140-22-7 | C13H14N4O | 242.282 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number ALF031390.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS525-05-3
IUPAC Namedisodium 3-hydroxy-4-nitrosonaphthalene-2,7-disulfonate
Molecular FormulaC10H5NNa2O8S2
InChI KeyDMKMTGULLYISBH-UHFFFAOYSA-L
SMILES[Na+].[Na+].OC1=C(N=O)C2=CC=C(C=C2C=C1S([O-])(=O)=O)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<5 % (1 g, 105°C)
Appearance (Form)Crystalline powder
Appearance (Color)Yellow to orange
Infrared spectrumConforms
Suitability(for Co determination) Passes test
1,5-Diphenylcarbazide is used in the colorimetric determination of chromium, osmium as well as in the detection of cadmium, mercury, magnesium, emetine and aldehydes.