1,2-Propanediol, ACS, 99.5% min
1,2-Propanediol, ACS, 99.5% min
1,2-Propanediol, ACS, 99.5% min
1,2-Propanediol, ACS, 99.5% min
1,2-Propanediol, ACS, 99.5% min
As a substitute for ethylene glycol and glycerol | CAS: 57-55-6 | C3H8O2 | 76.095 g/mol
Have Questions?
Catalog number ALF030948.AP
Price (MYR)
406.00
Add to cart
Request bulk or custom format
Price (MYR)
406.00
Add to cart
Specifications
Chemical Name or Material1, 2-Propanediol
CAS57-55-6
Melting Point°C to 60°C
Storage Note 1Hygroscopic
Recommended StorageAmbient temperatures; Store under Nitrogen
View more
1,2-Propanediol, ACS is used as a substitute for ethylene glycol and glycerol. It is a good solvent for resins. It dissolves many essential oils but is immiscible with fixed oils.