1,10-Phenanthroline monohydrate, ACS, Thermo Scientific Chemicals
1,10-Phenanthroline monohydrate, ACS, Thermo Scientific Chemicals
1,10-Phenanthroline monohydrate, ACS, Thermo Scientific Chemicals
1,10-Phenanthroline monohydrate, ACS, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,10-Phenanthroline monohydrate, ACS, Thermo Scientific Chemicals
CAS: 5144-89-8 | C12H10N2O | 198.23 g/mol
Have Questions?
Catalog NumberQuantity
ALF030910.0910 g
Catalog number ALF030910.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS619-58-9
IUPAC Name4-iodobenzoic acid
Molecular FormulaC7H5IO2
InChI KeyGHICCUXQJBDNRN-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)≥96.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)267.0-276.0?C
Appearance (Color)White to pale cream to pale orange to pale brown
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
1,10-Phenanthroline monohydrate is useful to study the DNA-protein interactions due to its nuclease activity. It is used as a redox indicator and reagent for spectrophotometric assay of silver. It acts as a chelating ligand for the determination of Fe, Pd and V. It also acts as a matrix metalloproteinase inhibitor. It is a photometric reagent for iron, which forms a red chelate with ferrous (iron II) ion.