(±)-Propylene oxide, >99%, Thermo Scientific Chemicals
(±)-Propylene oxide, >99%, Thermo Scientific Chemicals
(±)-Propylene oxide, >99%, Thermo Scientific Chemicals
(±)-Propylene oxide, >99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
(±)-Propylene oxide, >99%, Thermo Scientific Chemicals
CAS: 75-56-9 | C3H6O | 58.08 g/mol
Have Questions?
Catalog NumberQuantity
ALF030765.K21 L
Catalog number ALF030765.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS106-42-3
IUPAC Name1,4-xylene
Molecular FormulaC8H10
InChI KeyURLKBWYHVLBVBO-UHFFFAOYSA-N
SMILESCC1=CC=C(C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
Refractive Index1.4945-1.4975 @ 20?C
FormLiquid
Identification (FTIR)Conforms
Propylene oxide is used as a precursor in the production of polyether polyols, which is utilized for making polyurethane plastics, polypropylene glycols, propylene glycol ethers and propylene carbonate. It is involved in the preparation of biological samples for electron microscopy. It plays a vital role in the removal of residual ethanol previously used for dehydration. Further, it is used in thermobaric weapons, and microbial fumigation. It is also used as solvents, and in the preparation of surfactants, lubricants and in oil demulsifiers.