Anthrone, ACS, Thermo Scientific Chemicals
Anthrone, ACS, Thermo Scientific Chemicals
Anthrone, ACS, Thermo Scientific Chemicals
Anthrone, ACS, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Anthrone, ACS, Thermo Scientific Chemicals
Used for determination of carbohydrates
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF030739.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS24324-17-2
IUPAC Name(9H-fluoren-9-yl)methanol
Molecular FormulaC14H12O
InChI KeyXXSCONYSQQLHTH-UHFFFAOYSA-N
SMILESOCC1C2=CC=CC=C2C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
Melting Point (clear melt)102.0-108.0?C
Used for determination of carbohydrates Anthrone is used for the determination of carbohydrates. It is used as a general reagent in the colorimetric determination of carbohydrates in yeast and plant extracts. It is involved in organic synthesis. It is utilized for the determination of sugar in body fluids. It finds application as a laxative.