Anthrone, ACS, Thermo Scientific Chemicals
Anthrone, ACS, Thermo Scientific Chemicals
Anthrone, ACS, Thermo Scientific Chemicals
Anthrone, ACS, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Anthrone, ACS, Thermo Scientific Chemicals
Used for determination of carbohydrates
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number ALF030739.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS7758-95-4
IUPAC Nameλ²-lead(2+) dichloride
Molecular FormulaCl2Pb
InChI KeyHWSZZLVAJGOAAY-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Pb++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormBeads
Residual water< 100 ppm
Total Metal Impurities0.001% max.
Particle Size-10 Mesh
Used for determination of carbohydrates Anthrone is used for the determination of carbohydrates. It is used as a general reagent in the colorimetric determination of carbohydrates in yeast and plant extracts. It is involved in organic synthesis. It is utilized for the determination of sugar in body fluids. It finds application as a laxative.