Thioacetamide, ACS, 99% min, Thermo Scientific Chemicals
Thioacetamide, ACS, 99% min, Thermo Scientific Chemicals
Thioacetamide, ACS, 99% min, Thermo Scientific Chemicals
Thioacetamide, ACS, 99% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Thioacetamide, ACS, 99% min, Thermo Scientific Chemicals
CAS: 62-55-5 | C2H5NS | 75.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number ALF030638.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS611-99-4
IUPAC Name4-(4-hydroxybenzoyl)phenol
Molecular FormulaC13H10O3
InChI KeyRXNYJUSEXLAVNQ-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)C(=O)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Melting point217°C to 222°C
Titration with NaOH>=96.0 %
Appearance (Color)Off-white to beige
View more
Thioacetamide is used in qualitative inorganic analysis as an in-situ source for sulfide ions. It is used in analytical chemistry as a source of hydrogen sulfide. It is an additive in enantioselective reduction of beta-keto esters with immobilized bakers yeast. It is also used as a vulcanizing agent of polymer and a crosslinking agent.