1,10-Phenanthroline iron(II) sulfate, 0.025M aq. soln., Thermo Scientific Chemicals
1,10-Phenanthroline iron(II) sulfate, 0.025M aq. soln., Thermo Scientific Chemicals
1,10-Phenanthroline iron(II) sulfate, 0.025M aq. soln., Thermo Scientific Chemicals
1,10-Phenanthroline iron(II) sulfate, 0.025M aq. soln., Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,10-Phenanthroline iron(II) sulfate, 0.025M aq. soln., Thermo Scientific Chemicals
CAS: 14634-91-4 | C36H24FeN6O4S | 692.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number ALF030562.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS124655-17-0
IUPAC Name(2R)-2-{[(tert-butoxy)carbonyl]amino}-3,3-dimethylbutanoic acid
Molecular FormulaC11H21NO4
InChI KeyLRFZIPCTFBPFLX-JLDDOWRYNA-N
SMILESCC(C)(C)OC(=O)N[C@@H](C(O)=O)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
1,10-Phenanthroline iron(II) sulfate is used as an indicator for the iron phenanthroline compound in analytical chemistry. Further, it is used as a redox indicator for making oscillatory Belousov-Zhabotinsky reactions visible.