1,10-Phenanthroline iron(II) perchlorate, Thermo Scientific Chemicals
1,10-Phenanthroline iron(II) perchlorate, Thermo Scientific Chemicals
1,10-Phenanthroline iron(II) perchlorate, Thermo Scientific Chemicals
1,10-Phenanthroline iron(II) perchlorate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,10-Phenanthroline iron(II) perchlorate, Thermo Scientific Chemicals
Oxidation, reduction indicator
Have Questions?
Catalog NumberQuantity
ALF030557.0910 g
Catalog number ALF030557.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or Material1,10-Phenanthroline iron(II) perchlorate
CAS14586-54-0
Health Hazard 1H272-H315-H319-H335
Health Hazard 2GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
View more
1,10-Phenanthroline iron(II) perchlorate is used as an oxidation, reduction indicator. It is used for the detection, identification, analysis, of chemical, biological, or pathologic processes or conditions. Agents that are capable of inserting themselves between the successive bases in DNA, thus kinking, uncoiling or otherwise deforming it and therefore preventing its proper functioning. They are used in the study of DNA.