Peroxide, Quantofix™ Test Strips
Peroxide, Quantofix™ Test Strips
Peroxide, Quantofix™ Test Strips
Have Questions?
Catalog NumberQuantity
ALF023284.QA1 Kit
Catalog number ALF023284.QA
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 Kit
Request bulk or custom format
Chemical Identifiers
CAS336100-46-0
IUPAC Name1H-pyrrole-3-carboxylic acid
Molecular FormulaC5H5NO2
InChI KeyDOYOPBSXEIZLRE-UHFFFAOYSA-N
SMILESOC(=O)C1=CNC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige to brown
Titration with NaOH>=94 %
Infrared spectrumConforms
HPLC>=94 %
Appearance (Form)Crystalline powder