Phosphorus(III) chloride, 99.997% (metals basis), Thermo Scientific Chemicals
Phosphorus(III) chloride, 99.997% (metals basis), Thermo Scientific Chemicals
Phosphorus(III) chloride, 99.997% (metals basis), Thermo Scientific Chemicals
Phosphorus(III) chloride, 99.997% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphorus(III) chloride, 99.997% (metals basis), Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF022988.22100 g
Catalog number ALF022988.22
Price (MYR)
2,451.82
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
2,451.82
Add to cart
Specifications
Boiling Point76°C
Chemical Name or MaterialPhosphorus(III) chloride
Melting Point-111.8°C
CAS7719-12-2
Health Hazard 1H300+H330-H314-H373
View more
Phosphorus(III) chloride is the precursor to organ phosphorus compounds, most notably phosphites and phosphonates. It is used during electrode position of metal on rubber and for making surfactants, gasoline additives, dyestuffs, textile finishing agents, medicinal products, and other chemicals.