Dimethyl sulfide, 99+%, Thermo Scientific Chemicals
Dimethyl sulfide, 99+%, Thermo Scientific Chemicals
Dimethyl sulfide, 99+%, Thermo Scientific Chemicals
Dimethyl sulfide, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dimethyl sulfide, 99+%, Thermo Scientific Chemicals
CAS: 75-18-3 | C2H6S | 62.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Catalog number ALF022949.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialDimethyl sulfide
CAS75-18-3
Health Hazard 1H225-H319-H335
Health Hazard 2GHS H Statement
H225-H318-H302-H335-H315
Highly flammable liquid and vapor.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
View more
Dimethyl sulfide is used as a displaceable ligand in chloro(dimethyl sulfide)gold(I) and other coordination compounds, as a reducing agent in ozonolysis reactions as well as a food flavoring component. Further, it is utilized for the preparation of dimethyl sulfoxide, which is a common and important industrial solvent. It finds application in the petroleum industry.