Tetrahydrofuran, UV, HPLC Grade, 99.7+% min, unstabilized, Thermo Scientific Chemicals
Tetrahydrofuran, UV, HPLC Grade, 99.7+% min, unstabilized, Thermo Scientific Chemicals
Tetrahydrofuran, UV, HPLC Grade, 99.7+% min, unstabilized, Thermo Scientific Chemicals
Tetrahydrofuran, UV, HPLC Grade, 99.7+% min, unstabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetrahydrofuran, UV, HPLC Grade, 99.7+% min, unstabilized, Thermo Scientific Chemicals
CAS: 109-99-9 | C4H8O | 72.11 g/mol
Have Questions?
Catalog NumberQuantity
ALF022904.M14 x 1 L
Catalog number ALF022904.M1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 x 1 L
Request bulk or custom format
Chemical Identifiers
CAS553-86-6
IUPAC Name2,3-dihydro-1-benzofuran-2-one
Molecular FormulaC8H6O2
InChI KeyACZGCWSMSTYWDQ-UHFFFAOYSA-N
SMILESO=C1CC2=CC=CC=C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point47°C to 52°C
Appearance (Color)White to yellow to light orange
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Crystalline powder or crystals