2,6-Dimethylphenyl isocyanide, Thermo Scientific Chemicals
2,6-Dimethylphenyl isocyanide, Thermo Scientific Chemicals
2,6-Dimethylphenyl isocyanide, Thermo Scientific Chemicals
2,6-Dimethylphenyl isocyanide, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,6-Dimethylphenyl isocyanide, Thermo Scientific Chemicals
CAS: 2769-71-3 | C9H9N | 131.18 g/mol
Have Questions?
Catalog NumberQuantity
ALF020748.031 g
Catalog number ALF020748.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS39389-20-3
IUPAC Name1,2-diethenylbenzene; 2-ethenylbenzene-1-sulfonic acid
Molecular FormulaC18H18O3S
InChI KeySIWVGXQOXWGJCI-UHFFFAOYSA-N
SMILESC=CC1=CC=CC=C1C=C.OS(=O)(=O)C1=CC=CC=C1C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Opaque
Moisture holding capacity52 to 57% ( in the H form)
Exchange capacity>/= 1.7 meq/ml
FormBeads
2,6-Dimethylphenyl isocyanate is used in the preparation of homoleptic metallaamidine complex, derivatized bets-cyclodextrins. It is used as chiral stationary phase in normal-phase liquid chromatography. It is also involved in the preparation of tris( 2,6-dimethylphenylimido)methylrhenium (VII) and 1-(2-isopropylphenyl)-3-(2,6-dimethylphenyl)urea.