Potassium tricyanomethanide, Thermo Scientific Chemicals
Potassium tricyanomethanide, Thermo Scientific Chemicals
Potassium tricyanomethanide, Thermo Scientific Chemicals
Potassium tricyanomethanide, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Potassium tricyanomethanide, Thermo Scientific Chemicals
CAS: 34171-69-2 | C4KN3 | 129.163 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number ALF020702.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS6553-96-4
IUPAC Name2,4,6-tris(propan-2-yl)benzene-1-sulfonyl chloride
Molecular FormulaC15H23ClO2S
InChI KeyJAPYIBBSTJFDAK-UHFFFAOYSA-N
SMILESCC(C)C1=CC(C(C)C)=C(C(=C1)C(C)C)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline solid
Melting point93°C to 97°C
Appearance (Color)White to light beige
Infrared spectrumConforms
Titration Argentometric>=96.0 %
Potassium tricyanomethanide is a precursor used for the preparation of dicyano ketene aminals. It reacts with amines to prepare alkyl ammonium tricyanomethanide. Further, it is involved in the preparation of tricyanomethanide-bridged binuclear organometallic complexes such as Cp(dppe)FeC(CN)3 and Cp(PPh3)2RuC(CN)3. It is used as starting material utilized in the preparation of 5-Amino-3-bromo-1-(tert-butyl)-1H-pyrazole-4-carbonitrile, which undergoes sandmeyer reaction to get the corresponding diaminopyrazole, 5-amino-3-aryl-1-(tert-butyl)-1H-pyrazole-4-carboxamides.