Tetra-n-butylammonium phosphate, 99+%, Thermo Scientific Chemicals
Tetra-n-butylammonium phosphate, 99+%, Thermo Scientific Chemicals
Tetra-n-butylammonium phosphate, 99+%, Thermo Scientific Chemicals
Tetra-n-butylammonium phosphate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetra-n-butylammonium phosphate, 99+%, Thermo Scientific Chemicals
Used as a counter-ion in ion-pair chromatography of water-soluble vitamins
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
0.5 g
2 g
10 g
Catalog number ALF019261.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTetra-n-butylammonium phosphate
CAS5574-97-0
Health Hazard 1H500
Health Hazard 2GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
View more
Tetra-n-butylammonium phosphate is used as a counter-ion in ion-pair reagent in high-performance liquid chromatography for the simultaneous determination of chlorpheniramine and maleate of water-soluble vitamins. It acts as a phase transfer catalyst. Further, it is used as a buffering reagent in HPLC. In addition to this, it is used as a mobile phase additive in two-dimensional reversed-phase high-performance thin-layer chromatography for the separation of chenodeoxycholic acid and deoxycholic acid.