Tetra-n-butylammonium phosphate, 99+%, Thermo Scientific Chemicals
Tetra-n-butylammonium phosphate, 99+%, Thermo Scientific Chemicals
Tetra-n-butylammonium phosphate, 99+%, Thermo Scientific Chemicals
Tetra-n-butylammonium phosphate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Tetra-n-butylammonium phosphate, 99+%, Thermo Scientific Chemicals
Used as a counter-ion in ion-pair chromatography of water-soluble vitamins
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2 g
10 g
50 g
Catalog number ALF019261.02
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
0.5 g
Request bulk or custom format
Chemical Identifiers
CAS109-68-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.5% (sum of isomers)
FormLiquid
Refractive Index1.3795-1.3845 @ 20?C
Tetra-n-butylammonium phosphate is used as a counter-ion in ion-pair reagent in high-performance liquid chromatography for the simultaneous determination of chlorpheniramine and maleate of water-soluble vitamins. It acts as a phase transfer catalyst. Further, it is used as a buffering reagent in HPLC. In addition to this, it is used as a mobile phase additive in two-dimensional reversed-phase high-performance thin-layer chromatography for the separation of chenodeoxycholic acid and deoxycholic acid.