Nickel(II) 2,4-pentanedionate, 95%, Thermo Scientific Chemicals
Nickel(II) 2,4-pentanedionate, 95%, Thermo Scientific Chemicals
Nickel(II) 2,4-pentanedionate, 95%, Thermo Scientific Chemicals
Nickel(II) 2,4-pentanedionate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nickel(II) 2,4-pentanedionate, 95%, Thermo Scientific Chemicals
Catalyst and U.V. stabilizer
Have Questions?
Catalog NumberQuantity
ALF018811.1425 g
Catalog number ALF018811.14
Price (MYR)
394.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
394.00
Add to cart
Specifications
Chemical Name or MaterialNickel(II) 2,4-pentanedionate
CAS3264-82-2
Health Hazard 1H302-H317-H334-H350i-H500
Health Hazard 2GHS H Statement
H350-H302-H317
May cause cancer.
Harmful if swallowed.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P261-P264b-P270-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P341-P308+P313-P330-P333+P313-P342+P311-P363-P501c
View more
Catalyst and U.V. stabilizer.Nickel(II) 2,4-pentanedionate is used as a precursor to nickel bis(cyclooctadiene) catalyst. It is also used in the deposition of nickel(II) oxide thin film by sol-gel techniques on conductive glass substrates. Further, it is used in organic synthesis to produce organometals. It is associated with dimethylgold(III) acetylacetonate is used in gold on nickel plating.