Bis(2-ethylhexyl) phosphate, 95%, Thermo Scientific Chemicals
Bis(2-ethylhexyl) phosphate, 95%, Thermo Scientific Chemicals
Bis(2-ethylhexyl) phosphate, 95%, Thermo Scientific Chemicals
Bis(2-ethylhexyl) phosphate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bis(2-ethylhexyl) phosphate, 95%, Thermo Scientific Chemicals
Extraction of metals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2 kg
Catalog number ALF017723.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS26023-84-7
IUPAC Namedihydrogen hexachloroplatinumtetrakis(ylium)
Molecular FormulaCl6H2Pt
InChI KeyZKOQTCCVSRSECD-UHFFFAOYSA-J
SMILES[H+].[H+].Cl[Pt+4](Cl)(Cl)(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to orange-brown
Appearance (Form)Powder or crystals and/or chunks
Total trace metal impurities=<50 ppm
Assay>=37.0 % (Pt)
Extraction of metals.Bis(2-ethylhexyl) phosphate is used as a lubricant additive, corrosion inhibitor and a metal extractant. It is involved in the solvent extraction of uranium salts and rare earth metals. Iron(II) ions are extracted after reduction of Iron(III) ions. Further, it is used as a plasticizer and as a solvent in the synthesis of plastic.