Bis(2-ethylhexyl) phosphate, 95%, Thermo Scientific Chemicals
Bis(2-ethylhexyl) phosphate, 95%, Thermo Scientific Chemicals
Bis(2-ethylhexyl) phosphate, 95%, Thermo Scientific Chemicals
Bis(2-ethylhexyl) phosphate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bis(2-ethylhexyl) phosphate, 95%, Thermo Scientific Chemicals
Extraction of metals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number ALF017723.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS496-76-4
IUPAC Name5-hydroxy-1,2,3,4-tetrahydropyrimidine-2,4-dione
Molecular FormulaC4H4N2O3
InChI KeyOFJNVANOCZHTMW-UHFFFAOYSA-N
SMILESOC1=CNC(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)≥97.5%
FormPowder
Appearance (Color)White to pale cream
Extraction of metals.Bis(2-ethylhexyl) phosphate is used as a lubricant additive, corrosion inhibitor and a metal extractant. It is involved in the solvent extraction of uranium salts and rare earth metals. Iron(II) ions are extracted after reduction of Iron(III) ions. Further, it is used as a plasticizer and as a solvent in the synthesis of plastic.