Trimethylolpropane triacrylate, stabilized, Thermo Scientific Chemicals
Trimethylolpropane triacrylate, stabilized, Thermo Scientific Chemicals
Trimethylolpropane triacrylate, stabilized, Thermo Scientific Chemicals
Trimethylolpropane triacrylate, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trimethylolpropane triacrylate, stabilized, Thermo Scientific Chemicals
CAS: 15625-89-5 | C15H20O6 | 296.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ALF016445.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS13007-92-6
IUPAC Namehexakis(methanidylidyneoxidanium) chromium
Molecular FormulaC6CrO6
InChI KeyKOTQLLUQLXWWDK-UHFFFAOYSA-N
SMILES[Cr].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Appearance (Color)White to yellow
Infrared spectrumConforms
Elemental Analysis>=99.0 %