Benzo[a]pyrene, 96%, Thermo Scientific Chemicals
Benzo[a]pyrene, 96%, Thermo Scientific Chemicals
Benzo[a]pyrene, 96%, Thermo Scientific Chemicals
Benzo[a]pyrene, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzo[a]pyrene, 96%, Thermo Scientific Chemicals
Extensively used in cancer research
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
0.5 g
Catalog number ALF015856.MC
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mg
Request bulk or custom format
Chemical Identifiers
CAS105-58-8
IUPAC Namediethyl carbonate
Molecular FormulaC5H10O3
InChI KeyOIFBSDVPJOWBCH-UHFFFAOYSA-N
SMILESCCOC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥99.0%
Refractive Index1.3835-1.3855 @ 20?C
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
FormLiquid
Extensively used in cancer research and for GC (Gas Chromatography) and LC (Liquid Chromatography) Analysis. It is also a multipurpose intermediate.