Benzo[a]pyrene, 96%, Thermo Scientific Chemicals
Benzo[a]pyrene, 96%, Thermo Scientific Chemicals
Benzo[a]pyrene, 96%, Thermo Scientific Chemicals
Benzo[a]pyrene, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzo[a]pyrene, 96%, Thermo Scientific Chemicals
Extensively used in cancer research
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
100 mg
Catalog number ALF015856.02
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
0.5 g
Request bulk or custom format
Chemical Identifiers
CAS605-65-2
IUPAC Name5-(dimethylamino)naphthalene-1-sulfonyl chloride
Molecular FormulaC12H12ClNO2S
InChI KeyXPDXVDYUQZHFPV-UHFFFAOYSA-N
SMILESCN(C)C1=CC=CC2=C(C=CC=C12)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormPowder
Appearance (Color)Yellow to orange
Assay (HPLC)≥95.0%
Extensively used in cancer research and for GC (Gas Chromatography) and LC (Liquid Chromatography) Analysis. It is also a multipurpose intermediate.