Trioctyl phosphate, Thermo Scientific Chemicals
Trioctyl phosphate, Thermo Scientific Chemicals
Trioctyl phosphate, Thermo Scientific Chemicals
Trioctyl phosphate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trioctyl phosphate, Thermo Scientific Chemicals
CAS: 78-42-2 | C24H51O4P | 434.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number ALF015808.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS504-07-4
IUPAC Name1,3-diazinane-2,4-dione
Molecular FormulaC4H6N2O2
InChI KeyOIVLITBTBDPEFK-UHFFFAOYSA-N
SMILESO=C1CCNC(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder or lumps
Assay (HPLC)≥96.0%
Identification (FTIR)Conforms
Trioctyl phosphate finds application as a fire-retardant, plasticizer for polyvinyl chloride and cellulose nitrate. It is used as a stimulant for the chemical warfare nerve agent VX. Further, it used as an alkylation agent for nitrogen heterocyclic compounds and as a catalyst to produce phenolic and urea resins. In addition to this, it is used as an antifoaming agent and a co-solvent in hydrogen peroxide production.