Trioctyl phosphate, Thermo Scientific Chemicals
Trioctyl phosphate, Thermo Scientific Chemicals
Trioctyl phosphate, Thermo Scientific Chemicals
Trioctyl phosphate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trioctyl phosphate, Thermo Scientific Chemicals
CAS: 78-42-2 | C24H51O4P | 434.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number ALF015808.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS98-59-9
IUPAC Name4-methylbenzene-1-sulfonyl chloride
Molecular FormulaC7H7ClO2S
InChI KeyYYROPELSRYBVMQ-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)64.0-72.0?C
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Trioctyl phosphate finds application as a fire-retardant, plasticizer for polyvinyl chloride and cellulose nitrate. It is used as a stimulant for the chemical warfare nerve agent VX. Further, it used as an alkylation agent for nitrogen heterocyclic compounds and as a catalyst to produce phenolic and urea resins. In addition to this, it is used as an antifoaming agent and a co-solvent in hydrogen peroxide production.