Pentaerythritol triacrylate, stab. with 4-methoxyphenol
Pentaerythritol triacrylate, stab. with 4-methoxyphenol
Pentaerythritol triacrylate, stab. with 4-methoxyphenol
Pentaerythritol triacrylate, stab. with 4-methoxyphenol
Thermo Scientific Chemicals
Pentaerythritol triacrylate, stab. with 4-methoxyphenol
CAS: 3524-68-3 | C14H18O7 | 298.291 g/mol
Have Questions?
Catalog NumberdimQuantity
ALF015705.36
Catalog number ALF015705.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Formula Weight298.28
Unit Size
Pentaerythritol triacrylate is used in UV-cured inks, dental polymers and varnishes. Further, it is used in the preparation of printing inks, coatings, in photoresists, acrylic glues, adhesives, binders, paper and wood impregnates. In addition, it serves as a modifier for polyester and fiberglass and plastic components for the medical industry.