Copper(II) gluconate, Thermo Scientific Chemicals
Copper(II) gluconate, Thermo Scientific Chemicals
Copper(II) gluconate, Thermo Scientific Chemicals
Copper(II) gluconate, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Copper(II) gluconate, Thermo Scientific Chemicals
CAS: 527-09-3 | C12H22CuO14 | 453.84 g/mol
Have Questions?
Catalog NumberQuantity
ALF015325.1850 g
Catalog number ALF015325.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS646-30-0
IUPAC Namenonadecanoic acid
Molecular FormulaC19H38O2
InChI KeyISYWECDDZWTKFF-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.0 %
Infrared spectrumConforms
Appearance (Color)White
Appearance (Form)Glistening powder or flakes
Copper(II) gluconate is used in dietary supplement. It is also used to treat acne vulgaris, common cold, hypertension, premature labor, leishmaniasis and visceral postoperative complications. Further, it is used in oral deodorants and feed additives. In addition to this, it is used as a synergist and nutritional supplement for foods. It is the active component in retsyn used in mints and certs.