Gold foil, 0.25mm (0.01in) thick, 99.95% (metals basis), Thermo Scientific Chemicals
Gold foil, 0.25mm (0.01in) thick, 99.95% (metals basis), Thermo Scientific Chemicals
Gold foil, 0.25mm (0.01in) thick, 99.95% (metals basis), Thermo Scientific Chemicals
Gold foil, 0.25mm (0.01in) thick, 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gold foil, 0.25mm (0.01in) thick, 99.95% (metals basis), Thermo Scientific Chemicals
CAS: 7440-57-5 | Au | 196.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
25 x 25 mm
Catalog number ALF014722.FI
Price (MYR)
18,283.00
Add to cart
Quantity:
50 x 50 mm
Request bulk or custom format
Price (MYR)
18,283.00
Add to cart
Specifications
Chemical Name or MaterialGold foil
Name Note0.25mm (0.01 in.) thick
CAS7440-57-5
Recommended StorageAmbient temperatures
EINECS Number231-165-9