Manganese(II) oxalate dihydrate, Mn 30% min, Thermo Scientific Chemicals
Manganese(II) oxalate dihydrate, Mn 30% min, Thermo Scientific Chemicals
Manganese(II) oxalate dihydrate, Mn 30% min, Thermo Scientific Chemicals
Manganese(II) oxalate dihydrate, Mn 30% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Manganese(II) oxalate dihydrate, Mn 30% min, Thermo Scientific Chemicals
As a paint and varnish drier
Have Questions?
Catalog NumberQuantity
ALF014685.30250 g
Catalog number ALF014685.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS513-77-9
IUPAC Namebarium(2+) carbonate
Molecular FormulaCBaO3
InChI KeyAYJRCSIUFZENHW-UHFFFAOYSA-L
SMILES[Ba++].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Formpowder and/or chunks
Assay (special)?2.5 % of Theoretical value (Ba)
Total Metal Impurities≤0.05% (500 ppm)
Manganese(II) oxalate dihydrate is used as a paint and varnish drier. It is also used as a chemical reagent as well as to prepare other manganese compounds. In addition, it is used as a semiconductor photosensitive material.