Molybdenum wire, platinum coated, 0.457mm (0.018in) dia, Thermo Scientific Chemicals
Molybdenum wire, platinum coated, 0.457mm (0.018in) dia, Thermo Scientific Chemicals
Molybdenum wire, platinum coated, 0.457mm (0.018in) dia, Thermo Scientific Chemicals
Molybdenum wire, platinum coated, 0.457mm (0.018in) dia, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Molybdenum wire, platinum coated, 0.457mm (0.018in) dia, Thermo Scientific Chemicals
CAS: 7439-98-7 | Mo | 95.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 m
1 m
Catalog number ALF014658.G5
Price (MYR)
3,305.00
Add to cart
Quantity:
5 m
Request bulk or custom format
Price (MYR)
3,305.00
Add to cart
Specifications
Chemical Name or MaterialMolybdenum wire, platinum coated
Name Note0.457mm (0.018 in.) dia.
CAS7439-98-7
Recommended StorageAmbient temperatures
EINECS Number231-107-2