Bis(cyclopentadienyl)manganese, sublimed, Thermo Scientific Chemicals
Bis(cyclopentadienyl)manganese, sublimed, Thermo Scientific Chemicals
Bis(cyclopentadienyl)manganese, sublimed, Thermo Scientific Chemicals
Bis(cyclopentadienyl)manganese, sublimed, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bis(cyclopentadienyl)manganese, sublimed, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF014535.065 g
Catalog number ALF014535.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialBis(cyclopentadienyl)manganese
Name NoteSublimed
CAS73138-26-8
Health Hazard 1H228-H302+H312+H332-H315-H319-H335
Health Hazard 2GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only