Vanadium wire, 1.0mm (0.04 in.) dia., stress relieved, 99.7% (metals basis), Thermo Scientific Chemicals
Vanadium wire, 1.0mm (0.04 in.) dia., stress relieved, 99.7% (metals basis), Thermo Scientific Chemicals
Vanadium wire, 1.0mm (0.04 in.) dia., stress relieved, 99.7% (metals basis), Thermo Scientific Chemicals
Vanadium wire, 1.0mm (0.04 in.) dia., stress relieved, 99.7% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Vanadium wire, 1.0mm (0.04 in.) dia., stress relieved, 99.7% (metals basis), Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF014359.BY100 cm
Catalog number ALF014359.BY
Price (MYR)
1,572.00
Add to cart
Quantity:
100 cm
Request bulk or custom format
Price (MYR)
1,572.00
Add to cart
Specifications
Chemical Name or MaterialVanadium wire
Name Note1.0mm (0.04 in.) dia., Stress relieved
CAS7440-62-2
Recommended StorageAmbient temperatures
EINECS Number231-171-1