Barium, Oil based standard solution, Specpure™ Ba 1000μg/g
Barium, Oil based standard solution, Specpure™ Ba 1000μg/g
Thermo Scientific Chemicals
Barium, Oil based standard solution, Specpure™ Ba 1000μg/g
Have Questions?
Catalog NumberQuantity
ALF014230.1850 g
Catalog number ALF014230.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS51388-20-6
IUPAC Namehydrogen 4-(benzyloxy)aniline chloride
Molecular FormulaC13H14ClNO
InChI KeyKQBDLOVXZHOAJI-UHFFFAOYSA-N
SMILES[H+].[Cl-].NC1=CC=C(OCC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to brown or pale grey to pale pink to pink
FormCrystals or powder or crystalline powder
Assay (Argentometric Titration)≥96.0 to ≤104.0%