Boron, Oil based standard solution, Specpure™ B 1000μg/g
Boron, Oil based standard solution, Specpure™ B 1000μg/g
Thermo Scientific Chemicals
Boron, Oil based standard solution, Specpure™ B 1000μg/g
Have Questions?
Catalog NumberQuantity
ALF014227.1850 g
Catalog number ALF014227.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS7784-24-9
IUPAC Namealuminium(3+) potassium dodecahydrate disulfate
Molecular FormulaAlH24KO20S2
InChI KeyGNHOJBNSNUXZQA-UHFFFAOYSA-J
SMILESO.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[K+].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystals
Titration Complexometric98.0 to 102.0 %
Heavy metals (as Pb)=<10 ppm
Insoluble matter=<0.005 %