Magnesium, Oil based standard solution, Specpure™ Mg 1000μg/g
Magnesium, Oil based standard solution, Specpure™ Mg 1000μg/g
Magnesium, Oil based standard solution, Specpure™ Mg 1000μg/g
Magnesium, Oil based standard solution, Specpure™ Mg 1000μg/g
Thermo Scientific Chemicals
Magnesium, Oil based standard solution, Specpure™ Mg 1000μg/g
Have Questions?
Catalog NumberQuantity
ALF014219.1850 g
Catalog number ALF014219.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS123-62-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<20 APHA
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4030 to 1.4050 (20°C, 589 nm)