Potassium, Oil based standard solution, Specpure™ K 1000μg/g
Potassium, Oil based standard solution, Specpure™ K 1000μg/g
Potassium, Oil based standard solution, Specpure™ K 1000μg/g
Potassium, Oil based standard solution, Specpure™ K 1000μg/g
Thermo Scientific Chemicals
Potassium, Oil based standard solution, Specpure™ K 1000μg/g
Have Questions?
Catalog NumberQuantity
ALF014216.1850 g
Catalog number ALF014216.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS75-11-6
IUPAC Namediiodomethane
Molecular FormulaCH2I2
InChI KeyNZZFYRREKKOMAT-UHFFFAOYSA-N
SMILESICI
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to red to orange-brown