Molybdenum, Oil based standard solution, Specpure™ Mo 1000μg/g
Molybdenum, Oil based standard solution, Specpure™ Mo 1000μg/g
Thermo Scientific Chemicals
Molybdenum, Oil based standard solution, Specpure™ Mo 1000μg/g
Have Questions?
Catalog NumberQuantity
ALF014213.1850 g
Catalog number ALF014213.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS42330-59-6
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%